Skip to Content
Merck
All Photos(1)

Key Documents

GF52888146

Hastelloy® C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn

200mm tube, 6.35mm outside diameter, 0.9mm wall thickness, 4.55mm inside diameter

Synonym(s):

NI147200

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 528-881-46

L × wall thickness

200 mm × 0.9 mm

O.D. × I.D.

6.35 mm × 4.55 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Hastelloy is a registered trademark of Haynes International, Inc.
Hastelloy is a registered trademark of Haynes International, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service