GF43109917
Inconel® alloy 718 - heat resisting alloy, Ni53Fe19Cr19NbMoTi
rod, 200mm x 6.35mm diameter, annealed
Synonym(s):
NI137910
Select a Size
Vendor SKU: A111107-100MG
A$86.90
Availability
Select a Size
About This Item
Vendor SKU: A111107-100MG
A$86.90
Availability
Recommended Products
form
rod
manufacturer/tradename
Goodfellow 431-099-17
L × diam.
200 mm × 6.35 mm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service