Key Documents
BR474300
BRAND® transparent plastic staining trough with two lids
PMP, L × W × H 101 mm × 83 mm × 70 mm
Synonym(s):
Microscopy, Staining, Staining Accessories
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
material
PMP
manufacturer/tradename
BRAND 474300
L × W × H
101 mm × 83 mm × 70 mm
General description
The BRAND® transparent plastic staining trough is made from clear PMP, providing visibility for monitoring samples during staining processes. It features two lids: one designed to protect against evaporation when the trough is not in use, and another with a handle slot that facilitates easy handling during the staining process. This thoughtful design enhances usability and efficiency in laboratory settings, making it ideal for various staining applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)