BMSH3551180P
Benchmark ST180 PLUS CO2 Incuubator with three shelves
capacity 180 L, AC/DC input 115 V
Synonym(s):
CO2 Incubator, Humidity Control incubator, Lab incubator, Digital incubator, Laboratory Incubator, Tissue Culture Incubator, Cell Incubator, Cell Culture Incubator,
About This Item
Recommended Products
manufacturer/tradename
Benchmark Scientific H3551-180P
parameter
temp. range at Ambient +5 to 60 °C
W × L × H
47 cm × 52 cm × 71 cm, Internal dimensions
capacity
180 L
Looking for similar products? Visit Product Comparison Guide
General description
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service