Key Documents
BAF182020000
SP Bel-Art® Chromatography Column Holder
diam. × H 12.25 in. (311 mm) × 2.5 in. (64 mm)
Synonym(s):
column holder
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18202-0000
diam. × H
12.25 in. (311 mm) × 2.5 in. (64 mm)
General description
Securely holds as many as eight chromatography columns up to 30mm (1³⁄₁₆") in diameter in or between its flexible loops.
Features and Benefits
  • Rigid plastic back is mounted on two polyethylene support blocks which slide over support stand rods
  • Tighten the screws to mount the rack at the desired height
  • Requires two support rods and stands (H18303-0000 sold separately) or can be wall-mounted by removing the support blocks from the back of the unit
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)