Skip to Content
Merck
All Photos(1)

Key Documents

750476

Sigma-Aldrich

6-Hydroxyhexanoic acid-2,2,3,3,4,4,5,5,6,6-d10

95 atom % D, 97% (CP)

Synonym(s):

6-Hydroxycaproic acid-2,2,3,3,4,4,5,5,6,6-d10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CD2)5COOH
Molecular Weight:
142.22
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

isotopic purity

95 atom % D

Assay

97% (CP)

form

solid

mass shift

M+10

storage temp.

−20°C

SMILES string

[2H]C([2H])(O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C6H12O3/c7-5-3-1-2-4-6(8)9/h7H,1-5H2,(H,8,9)/i1D2,2D2,3D2,4D2,5D2

InChI key

IWHLYPDWHHPVAA-YXALHFAPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service