375772
1-Acetyl-3-indolecarboxaldehyde
98%
Synonym(s):
N-Acetylindol-3-carboxaldehyde, NSC 61289
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
mp
165 °C (lit.)
functional group
aldehyde
SMILES string
CC(=O)n1cc(C=O)c2ccccc12
InChI
1S/C11H9NO2/c1-8(14)12-6-9(7-13)10-4-2-3-5-11(10)12/h2-7H,1H3
InChI key
LCJLFGSKHBDOAY-UHFFFAOYSA-N
General description
1-Acetyl-3-indolecarboxaldehyde participates in the preparation and characterization of three RNA-specific fluorescent probes (E36, E144 and F22), useful in live cell imaging.
Application
1-Acetyl-3-indolecarboxaldehyde may be used to prepare the thiosemicarbazones, via condensation reaction.
Reactant for synthesis of α-ketoamides as inhibitors of Dengue virus protease with antiviral activity in cell-culture
Reactant for preparation of homoallylic amines as antimicrobial agents
Reactant for preparation of pyrrole-based hydrazones as potential tuberculostatics
Reactant for synthesis of neoechinulin A and derivatives
Reactant for synthesis of substituted (Z)-2-(N-benzylindol-3-ylmethylene)quinuclidin-3-one and (Z)-(±)-2-(N-benzylindol-3-ylmethylene)quinuclidin-3-ol derivatives as potent thermal sensitizing agents
Reactant for preparation of RNA-specific live cell imaging probes E36, E144 and F22
Reactant for preparation of homoallylic amines as antimicrobial agents
Reactant for preparation of pyrrole-based hydrazones as potential tuberculostatics
Reactant for synthesis of neoechinulin A and derivatives
Reactant for synthesis of substituted (Z)-2-(N-benzylindol-3-ylmethylene)quinuclidin-3-one and (Z)-(±)-2-(N-benzylindol-3-ylmethylene)quinuclidin-3-ol derivatives as potent thermal sensitizing agents
Reactant for preparation of RNA-specific live cell imaging probes E36, E144 and F22
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis, Characterization, and Crystal Structure Analysis of the First Terdentate [C, N, S] Thiosemicarbazone Complex with a Six-Membered Palladacycle: Influence of Steric Effects on Ring Size.
European Journal of Inorganic Chemistry, 15, 3016-3021 (2006)
Nature protocols, 1(6), 2922-2932 (2007-04-05)
This protocol outlines a methodology for the preparation and characterization of three RNA-specific fluorescent probes (E36, E144 and F22) and their use in live cell imaging. It describes a detailed procedure for their chemical synthesis and purification; serial product characterization
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service