Skip to Content
Merck
All Photos(1)

Key Documents

SRP2078

Sigma-Aldrich

p53 (1-342) C-terminal deletion human

recombinant, expressed in insect cells, ≥80% (SDS-PAGE)

Synonym(s):

FLJ92943, LFS1, P53, TRP53

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in insect cells

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~39.5 kDa

packaging

pkg of 5 μg

concentration

250 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TP53(7157)

Biochem/physiol Actions

Human p53 protein is composed of 393 amino acid residues with several distinct regions. The N-terminal activation domain allows p53 protein to recruit the basal transcription machinery and activate the expression of target genes. The core domain binds to target DNA in a sequence-specific manner and the majority of mutations found in human tumors occur in the region of the gene encoding this domain. The C-terminal domain is composed of predominantly basic residues and modification of the C-terminal basic domain, including acetylation, glycosylation and phosphorylation, is an essential mechanism for regulating p53 function. Disruption or loss of oligomerization function is associated with loss of cell cycle arrest. This mutant protein (with the deletion of the C-terminus 51 residues including the entire basic domain and a portion of the tetramerization domain) can be used as a unique tool to study specific functions of p53 related to the C-terminus.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Chen et al.
Genes & development, 10(19), 2438-2451 (1996-10-01)
It is well established that induction of the p53 tumor suppressor protein in cells can lead to either cell cycle arrest or apoptosis. To further understand features of p53 that contribute to these cell responses several p53-null Saos2 and H1299
M Hollstein et al.
Science (New York, N.Y.), 253(5015), 49-53 (1991-07-05)
Mutations in the evolutionarily conserved codons of the p53 tumor suppressor gene are common in diverse types of human cancer. The p53 mutational spectrum differs among cancers of the colon, lung, esophagus, breast, liver, brain, reticuloendothelial tissues, and hemopoietic tissues.
W S el-Deiry et al.
Nature genetics, 1(1), 45-49 (1992-04-01)
Recent experiments have suggested that p53 action may be mediated through its interaction with DNA. We have now identified 18 human genomic clones that bind to p53 in vitro. Precise mapping of the binding sequences within these clones revealed a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service