Skip to Content
Merck
All Photos(1)

Key Documents

SAB2100641

Sigma-Aldrich

Anti-EARS2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Glutamyl-tRNA synthetase 2, mitochondrial (putative), Anti-KIAA1970, Anti-MSE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human, rat
citations:
2
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

59 kDa

species reactivity

human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EARS2(124454)

General description

The previously assigned protein identifier Q86YH3 has been merged into Q5JPH6. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the middle region of human EARS2

Biochem/physiol Actions

EARS2 belongs to the class-I aminoacyl-tRNA synthetase family. The function of the EARS2 protein is not known.

Sequence

Synthetic peptide located within the following region: TAKHLLLYQALGWQPPHFAHLPLLLNRDGSKLSKRQGDVFLEHFAADGFL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Late-onset leukoencephalopathy in a patient with recessive EARS2 mutations.
Edoardo Monfrini et al.
Neurology. Genetics, 6(5), e488-e488 (2020-08-18)
Nathan McNeill et al.
Neurology. Genetics, 3(4), e162-e162 (2017-07-28)
To investigate the genetic etiology of a patient diagnosed with leukoencephalopathy, brain calcifications, and cysts (LCC). Whole-exome sequencing was performed on a patient with LCC and his unaffected family members. The variants were subject to in silico and in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service