Skip to Content
Merck
All Photos(3)

Key Documents

L3510

Sigma-Aldrich

Lucifer Yellow VS dilithium salt

≥80% purity, powder

Synonym(s):

4-Amino-N-(3-[vinylsulfonyl]phenyl)naphthalimide-3,6-disulfonate dilithium salt, Dilithium 4-amino-N-[3-(vinylsulfonyl)phenyl]naphthalimide-3,6-disulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12Li2N2O10S3
CAS Number:
Molecular Weight:
550.39
Beilstein:
5226753
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Lucifer Yellow VS dilithium salt, ~85%

Assay

~85%

form

powder

color

yellow to brown

fluorescence

λex 428 nm; λem 536 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Li+].[Li+].Nc1c(cc2C(=O)N(c3cccc(c3)S(=O)(=O)C=C)C(=O)c4cc(cc1c24)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C20H14N2O10S3.2Li/c1-2-33(25,26)11-5-3-4-10(6-11)22-19(23)14-8-12(34(27,28)29)7-13-17(14)15(20(22)24)9-16(18(13)21)35(30,31)32;;/h2-9H,1,21H2,(H,27,28,29)(H,30,31,32);;/q;2*+1/p-2

InChI key

PSJNJVOFDDMXDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ludĕk Bláha et al.
Toxicon : official journal of the International Society on Toxinology, 55(1), 126-134 (2009-07-22)
Toxicity and liver tumor promotion of cyanotoxins microcystins have been extensively studied. However, recent studies document that other metabolites present in the complex cyanobacterial water blooms may also have adverse health effects. In this study we used rat liver epithelial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service