Skip to Content
Merck
All Photos(1)

Key Documents

W455530

Sigma-Aldrich

Dihydrogalangal acetate

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16O4
CAS Number:
Molecular Weight:
236.26
FEMA Number:
4555
UNSPSC Code:
12164502
Flavis number:
09.946
NACRES:
NA.21

grade

FG

Quality Level

reg. compliance

EU 1334/2008& 178/2002
FDA 21 CFR 117

Assay

≥95%

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

pungent; spicy

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service