Skip to Content
Merck
All Photos(1)

Key Documents

BCR152

Dibenz[a,i]acridine

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H13N
CAS Number:
Molecular Weight:
279.33
Beilstein:
209225
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2cc3nc4ccc5ccccc5c4cc3cc2c1

InChI

1S/C21H13N/c1-2-7-16-13-21-17(11-15(16)6-1)12-19-18-8-4-3-5-14(18)9-10-20(19)22-21/h1-13H

InChI key

MRXMZJQYYWLOCW-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR152

Legal Information

BCR is a registered trademark of European Commission

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service