Key Documents
58384
Hamilton® syringe, 700 series, special needle
725SNR, volume 250 μL, needle size 22 ga (blunt tip), needle L 51 mm (2 in.)
About This Item
UNSPSC Code:
41122001
Pricing and availability is not currently available.
material
stainless steel needle (point style 3)
needle size
22 ga (blunt tip)
feature
needle type SN (special needle)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80765
needle L
51 mm (2 in.)
volume
250 μL
compatibility
for sampling Valco VISF-2
for use with Rheodyne
for use with SSI
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)