Skip to Content
Merck
All Photos(1)

Key Documents

50632

Supelco

2-Dimethylaminoethanol

analytical standard

Synonym(s):

N,N-Dimethyl-2-hydroxyethylamine, N,N-Dimethylethanolamine, Jeffcat DMEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
1209235
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

100 mmHg ( 55 °C)
6.12 mmHg ( 20 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.1% water

refractive index

n20/D 1.4294 (lit.)

bp

134-136 °C (lit.)

mp

−70 °C (lit.)

solubility

H2O: soluble

density

0.886 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical

format

neat

SMILES string

CN(C)CCO

InChI

1S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3

InChI key

UEEJHVSXFDXPFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - Seta closed cup

Flash Point(C)

40 °C - Seta closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

2-Dimethylaminoethanol (deanol): a brief review of its clinical efficacy and postulated mechanism of action.
O Re'
Current therapeutic research, clinical and experimental, 16(11), 1238-1242 (1974-11-01)
Tanja Peters et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 94, 141-146 (2014-09-06)
The [(18)F]fluoroethyl moiety has been widely utilized in the synthesis of (18)F-labelled compounds. The aim of this work was the reliable synthesis of [(18)F]FEtOTf with a novel strategy to increase the reactivity of the commonly used [(18)F]FEB and [(18)F]FEtOTos. [(18)F]FEtOTf
Meriyem Aktas et al.
The FEBS journal, 281(15), 3523-3541 (2014-06-17)
Phosphatidylcholine (PC) is a rare membrane lipid in bacteria, but is crucial for virulence of the plant pathogen Agrobacterium tumefaciens and various other pathogens. Agrobacterium tumefaciens uses two independent PC biosynthesis pathways. One is dependent on the integral membrane protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service