Skip to Content
Merck
All Photos(1)

Key Documents

PLGCD08205

Millipore

Ultracel® Ultrafiltration Membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32161003
NACRES:
NB.24

material

composite regenerated cellulose (CRC)

Quality Level

grade

for separation

description

Filter code PLGCD

technique(s)

ultrafiltration: suitable

filtration area

20 cm2

membrane size

20 cm × 20 cm

matrix

Ultracel®

pore size

8 kDa filter

Legal Information

ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service