Z219738
Aldrich® manifold with trap
male joint: ST/NS 14/20 (1), male Joints: ST/NS 24/40 (2), PTFE valves
All Photos(1)
About This Item
UNSPSC Code:
40161501
NACRES:
NB.79
Recommended Products
Joint
male Joints: ST/NS24/40 (2)
male joint: ST/NS14/20 (1)
feature
1 hose barb connection
Related Categories
General description
Features four 0-10 mm bore high-vacuum PTFE valves and a removable end cap for easy cleaning. Approx. 580 mm L. In-line trap eliminates extra hookup, thus improving vacuum performance.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)