Key Documents
SYNF701430M
Synthware fritted filter tube
joint: ST/NS 14/20, disc O.D. 30 mm, porosity: medium
Synonym(s):
Synthware F701430M, straight filter tube
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F701430M
disc O.D.
30 mm
General description
Available with a choice of coarse or medium porosity fritted discs.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)