Skip to Content
Merck
All Photos(2)

Documents

DWK651486

Wheaton® Cryule Cryogenic Ampule

Synonym(s):

651486

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.17

feature

preconditioned (pre-scored)

packaging

pkg of 144 ea

manufacturer/tradename

WHEATON® 651486

capacity

2 mL

diam. × H

11.5 mm × 70 mm

Looking for similar products? Visit Product Comparison Guide

Application

Biological Sample Preservation

Features and Benefits

  • For biological sample preservation in glass
  • Pre-scored to eliminate filing
  • Allows for storage at low temperatures
  • Can be used in the vapor phase of liquid nitrogen
  • Special design allows for storage at low temperatures as well as rapid thawing
  • Converted from Type I borosilicate glass tubing, ampules conform to the hydrolytic resistance requirements outlined in USP and EP 3.2.1.

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service