444111
Erbium(III) perchlorate solution
40 wt. % in H2O, 99.9% trace metals basis
Synonym(s):
Erbium triperchlorate
About This Item
Recommended Products
grade
for analytical purposes
Quality Level
Assay
99.9% trace metals basis
form
liquid
reaction suitability
reagent type: catalyst
core: erbium
concentration
40 wt. % in H2O
impurities
≤1500.0 ppm Trace Rare Earth Analysis
density
1.44 g/mL at 25 °C
SMILES string
[Er+3].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O
InChI
1S/3ClHO4.Er/c3*2-1(3,4)5;/h3*(H,2,3,4,5);/q;;;+3/p-3
InChI key
FGBKFKCLWFRKEW-UHFFFAOYSA-K
Related Categories
Application
- To synthesize luminescent metal-organicframeworks.
- As a rare-earth dopant along with an organicprecursor to prepare fluorophosphate glasses.
- As a startingmaterial in the preparation of [Er2(Gly)6(H2O)4](ClO4)6 .5H2O,a rare earth complex coordinated with amino acid.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Ox. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service