- Cysteinesulfonate and beta-sulfopyruvate metabolism. Partitioning between decarboxylation, transamination, and reduction pathways.
Cysteinesulfonate and beta-sulfopyruvate metabolism. Partitioning between decarboxylation, transamination, and reduction pathways.
L-Cysteinesulfonate (L-cysteate) is present in plasma, urine, and tissues in concentrations comparable to that of L-cysteinesulfinate, the primary oxidative metabolite of L-cysteine. Although cysteinesulfonate is known to be decarboxylated to taurine by cysteinesulfinate decarboxylase, the occurrence and importance of other metabolisms has not been examined. The present studies indicate that cysteinesulfonate partitions in vivo between decarboxylation and transamination; the latter reaction is catalyzed by aspartate aminotransferase and yields beta-sulfopyruvate. Whereas beta-sulfinylpyruvate, the product of cysteinesulfinate transamination, decomposes spontaneously, beta-sulfopyruvate is stable and is reduced by malate dehydrogenase to beta-sulfolactate. When L-[1-14C]cysteinesulfonate is given to mice, 60-75% is decarboxylated to taurine and about 25% is excreted in the urine as beta-sulfolactate. beta-Sulfo[1-14C] pyruvate is found to partition about equally between beta-sulfolactate and cysteinesulfonate formation; greater than 90% of the latter is decarboxylated. Parenterally administered beta-sulfo[1-14C]lactate is mostly excreted in the urine, but 12% is metabolized via beta-sulfopyruvate and cysteinesulfonate to 14CO2 and taurine. beta-Sulfopyruvate is not excreted, and only traces of sulfoacetate, perhaps formed by oxidative decarboxylation, are detected. These studies establish that cysteinesulfonate, beta-sulfopyruvate, and beta-sulfolactate are reversibly interconverted in vivo. Since only cysteinesulfonate is directly metabolized to CO2, the rate of 14CO2 formation from L-[1-14C]cysteinesulfonate is a valid measure of total cysteinesulfinate decarboxylase activity in vivo; use of this assay permits inhibitor effects to be accurately determined in intact mice. Thus, whereas in vitro assays indicate that beta-methyleneaspartate inhibits brain, liver, and kidney cysteinesulfinate decarboxylase by 0, greater than 60, and 90%, respectively, in vivo studies with L-[1-14C]cysteinesulfonate show net metabolic inhibition is about 40%.