Merck
  • On benzo[b][1,4]diazepinium-olates, -thiolates and -carboxylates as anti-Hückel mesomeric betaines.
On benzo[b][1,4]diazepinium-olates, -thiolates and -carboxylates as anti-Hückel mesomeric betaines.
Organic & biomolecular chemistry (2003-12-20)
Andreas Schmidt, Abbas Gholipour Shilabin, Martin Nieger
ABSTRACT
2,3-Diaminophenol 4, 3,4-diaminophenol 5, 4-methoxy-1,2-diaminobenzene 6, 3,4-diaminobenzenethiol 7, 2,3-diaminobenzoic acid 8, and 3,4-diaminobenzoic acid 9 were reacted with 2,4-pentanedione to yield the corresponding benzo[b][1,4]diazepinium salts, respectively. The hydroxy-benzo[b][1,4]diazepinium salts 17 and 18 do not form mesomeric betaines (MB) on deprotonation. Instead, they are converted into the diimines 24 and 25. By contrast, the 7-mercaptobenzo[b][1,4]diazepinium salt 20 yields the corresponding thiolate on increasing the pH of the solution. This MB, which possesses 4n pi-electrons, does not fit into the classification system of heterocyclic mesomeric betaines accepted today. Deprotonation of the betaine results in the formation of an instable anionic thiolate 31 which oxidizes immediately to the disulfide 32. The carboxy derivatives 21 and 22 readily form cross-conjugated mesomeric betaines. Whereas the diimine 34 proved to be instable, the sodium salt of the diimine 36 was unambiguously characterized. An X-ray single crystal analysis of 22 as its picrate is presented in order to gain additional insights into these 4n pi-electron systems.
Sigma-Aldrich
2,3-Diaminophenol, 97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
100 μL
In Stock
Details...
$515.00
$463.50
200 μL
In Stock
Details...
$678.00
To order products, please contact your local dealer.