Merck
  • Development of a gas chromatographic-mass spectrometric drug screening method for the N-dealkylated metabolites of fentanyl, sufentanil, and alfentanil.
Development of a gas chromatographic-mass spectrometric drug screening method for the N-dealkylated metabolites of fentanyl, sufentanil, and alfentanil.
Journal of chromatographic science (1997-10-23)
A K Valaer, T Huber, S V Andurkar, C R Clark, J DeRuiter
ABSTRACT
A sensitive, specific urinary assay for fentanyl, sufentanil, and alfentanil based on their N-dealkylated metabolites is described. Norfentanyl, norsufentanil-noralfentanil, and 2H5-norfentanyl are synthesized and characterized by standard analytical techniques. Derivatization of these secondary amines to yield the pentafluorobenzamides produces stable products with good gas chromatographic properties and unique, high-mass fragments in their mass spectra. These properties are utilized to develop a drug screening procedure based on gas chromatography-mass spectrometry to detect these major metabolites in human urine. The metabolites are isolated from urine samples by a liquid-liquid extraction procedure. The method allows for detection of metabolite concentrations as low as 0.3 ng/mL.
Product Description
Supelco
Norfentanyl oxalate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.