Merck
  • [Study of interneuronal connections by the technic of luminescent demonstration of retrograde transport of primulin].
[Study of interneuronal connections by the technic of luminescent demonstration of retrograde transport of primulin].
Arkhiv anatomii, gistologii i embriologii (1981-01-01)
V A Otellin, V I Meshkenaĭte, V L Rybakov
ABSTRACT
A possibility to study associative and projective connections of the optic cortical fields by the method of luminescent revealing of primuline retrograde transport has been demonstrated in the experiments with cats. Initial neurons of the connective systems have been revealed in various fields of the optic cortex (fields 17, 18, 19) and in the lateral geniculate body of the thalamus. The initial cells have various luminescent intensity.
Product Description
Supelco
Malvidin-3-galactoside chloride, analytical standard