Merck
  • Anti-obesity effects of glabridin-rich supercritical carbon dioxide extract of licorice in high-fat-fed obese mice.
Anti-obesity effects of glabridin-rich supercritical carbon dioxide extract of licorice in high-fat-fed obese mice.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-09-13)
Jiyun Ahn, Hyunjung Lee, Jayoung Jang, Suna Kim, Taeyoul Ha
ABSTRACT
Licorice (Glycyrrhiza glabra Linne) is a well-known medicinal plant and glabridin is an isoflavan isolated from licorice. In this study, we investigated the anti-obesity effect of glabridin and glabridin-rich supercritical fluid extract of licorice (LSC). Glabridin effectively inhibited adipogenesis in 3T3-L1 cells. Moreover, LSC showed inhibitory effect on adipogenesis in a dose-dependent manner. The inhibitory effect of LSC resulted from inhibiting the induction of the transcriptional factors CCAAT enhancer binding protein alpha and peroxisome proliferator-activated receptor gamma. Then we fed mice with high-fat diet containing none, 0.1% and 0.25% LSC for 8weeks to explore the anti-obesity effect of LSC in vivo. LSC significantly reduced weight gain by high-fat diet in a dose-dependent manner. The reductions of the hypertrophy of white adipose tissue and of fat cell size were also observed. In the liver, LSC supplementation effectively inhibited high-fat diet-induced hepatic steatosis through downregulation of gluconeogenesis related phosphoenolpyruvate carboxykinase and glucose 6-phosphatase and upregulation of the β-oxidation related carnitine palmitoyltransferase 1. Taken together, our results suggest that glabridin and glabridin-rich licorice extract would be effective anti-obesity agents.
Sigma-Aldrich
Glabridin, ≥98% (HPLC), powder