Merck
  • Influence of conformationally constrained amino acids replacing positions 2 and 3 of arginine vasopressin (AVP) and its analogues on their pharmacological properties.
Influence of conformationally constrained amino acids replacing positions 2 and 3 of arginine vasopressin (AVP) and its analogues on their pharmacological properties.
Protein and peptide letters (2007-03-10)
Dariusz Sobolewski, Adam Prahl, Izabela Derdowska, Anna Kwiatkowska, Jirina Slaninová, Bernard Lammek
ABSTRACT
Synthesis of thirteen new analogues of arginine vasopressin (AVP) has been described. Amino acid residues at positions 2 and 3 of AVP, [3-mercaptopropionic acid (Mpa)(1)]AVP (dAVP), [Mpa(1),d-Arg(8)]VP (dDAVP) and [Mpa(1),Val(4),d-Arg(8)]VP (dVDAVP) were replaced with one amino acid residue using sterically constrained non-proteinogenic amino acids, 4-aminobenzoic acid (Abz), cis-4-aminocyclohexanecarboxylic acid (ach) or its trans-isomer (Ach). In the case of a potent V(1a) antagonist, [1-mercaptocyclohexaneacetic acid (Cpa)(1)]AVP, only one similar analogue has been prepared by replacing positions 2 and 3 with Abz. Unfortunately, all new peptides were inactive in bioassays for the pressor, antidiuretic and uterotonic in vitro activities in the rat.