Merck
  • ER beta: identification and characterization of a novel human estrogen receptor.
ER beta: identification and characterization of a novel human estrogen receptor.
FEBS letters (1996-08-19)
S Mosselman, J Polman, R Dijkema
ABSTRACT
A novel estrogen receptor (hereinafter referred to as ER beta) was cloned using degenerate PCR primers. A comparison of the amino acid sequence of ER beta with the "classical' ER (ER alpha) shows a high degree of conservation of the DNA-binding domain (96%), and of the ligand-binding domain (58%). In contrast, the A/B domain, the hinge region and the F-domain are not conserved. Northern blot analysis revealed that ER beta is expressed in human thymus, spleen, ovary and testis. Transient transfections of an ER beta expression construct together with an ERE-based reporter construct in CHO cells clearly demonstrated transactivation of ER beta by 17 beta-estradiol. In addition, the ER alpha antagonist ICI-164384 is a potent antagonist for ER beta as well. Interestingly, the level of transactivation by 17 beta-estradiol is higher for ER alpha than for ER beta, which may reflect suboptimal conditions for ER beta at the level of the ligand, responsive element or cellular context.
Sigma-Aldrich
Estrogen Receptor-α human, ≥80% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution
Pricing and availability is not currently available.