MilliporeSigma
  • Mouse models for peroxisome biogenesis defects and β-oxidation enzyme deficiencies.
Mouse models for peroxisome biogenesis defects and β-oxidation enzyme deficiencies.
Biochimica et biophysica acta (2012-03-27)
Myriam Baes, Paul P Van Veldhoven
ABSTRACT
Peroxisome biogenesis and peroxisomal β-oxidation defects are rare inherited metabolic disorders in which several organs can be affected. A panel of mouse models has been created in which genes crucial to these processes were inactivated and the ensuing pathologies studied. In mice with enzyme defects of peroxisomal β-oxidation, the disease state strongly depends on the kind of substrates that are metabolized by the enzyme and the dietary composition. Because mice with generalized biogenesis defects seldom reach adulthood, conditional knockout models were generated to study the consequences of peroxisome deficiency in hepatocytes, different brain cell types and Sertoli cells. Although the precise relationship between the biochemical anomalies and pathologies was often not resolved, the mouse models allowed to document in detail histological abnormalities, metabolic and gene expression deregulations some of which are mediated by PPARα, and to uncover the essential role of peroxisomes in some unsuspected cell types.