Merck
  • The natural polyamine spermine functions directly as a free radical scavenger.
The natural polyamine spermine functions directly as a free radical scavenger.
Proceedings of the National Academy of Sciences of the United States of America (1998-09-16)
H C Ha, N S Sirisoma, P Kuppusamy, J L Zweier, P M Woster, R A Casero
ABSTRACT
The polyamines are small organic cations that are absolutely required for eukaryotic cell growth. Although their growth requirements are well established, the molecular functions of the polyamines are ill-defined. Oxidative damage to DNA by reactive oxygen species is a continual problem that cells must guard against to survive. The polyamine spermine, which is normally found in millimolar concentrations in the nucleus, is shown here to function directly as a free radical scavenger, and adducts formed as a result of this function are identified. These data suggest that spermine is a major natural intracellular compound capable of protecting DNA from free radical attack.
Sigma-Aldrich
Spermine tetrahydrochloride, powder or crystals
Pricing and availability is not currently available.
Sigma-Aldrich
Spermine, ≥97%
Pricing and availability is not currently available.