Merck
Stabilization of bovine intestine alkaline phosphatase by sugars.
Bioscience, biotechnology, and biochemistry (2012-01-11)
Satoshi Sekiguchi, Yasuhiko Hashida, Kiyoshi Yasukawa, Kuniyo Inouye
ABSTRACT
Bovine intestine alkaline phosphatase (BIALP) is widely used as a signaling enzyme in sensitive assays such as enzyme immunoassay. In this study, we evaluated the effects of sugars on the kinetic stability of BIALP in the hydrolysis of p-nitrophenylphosphate (pNPP). The temperatures reducing initial activity by 50% in a 30-min incubation, T(50), of BIALP with 1.0 M disaccharide (sucrose and trehalose) or 2.0 M monosaccharide (glucose and fructose) were 55.0-55.5 °C, 4.7-5.2 °C higher than without sugar (50.3±0.1 °C). The T(50) of BIALP increased to 58.4±0.3 °C when the trehalose concentration was from 1.0 to 1.5 M, but did not change when the glucose concentration was from 2.0 to 3.0 M. Thermodynamic analysis revealed that the stabilization of BIALP by sugars was driven by the increase in the enthalpy change of activation for thermal inactivation of BIALP. No sugars affected the k(cat) of BIALP in the hydrolysis of pNPP. These results suggest that not only trehalose, which is considered the most effective stabilizer of enzymes, but also sucrose, glucose, and fructose can be used as stabilizers of BIALP.