Merck
  • Tempol protects against oxidative damage and delays epithelial tumor onset in Fanconi anemia mice.
Tempol protects against oxidative damage and delays epithelial tumor onset in Fanconi anemia mice.
Cancer research (2008-03-05)
Qing-Shuo Zhang, Laura Eaton, Eric R Snyder, Scott Houghtaling, James B Mitchell, Milton Finegold, Carter Van Waes, Markus Grompe
ABSTRACT
Fanconi anemia (FA) is a genetic disorder characterized by congenital abnormalities, bone marrow failure, and marked cancer susceptibility. FA patients have an elevated risk of developing hematologic malignancies and solid tumors. Using Fancd2(-/-) knockout mice as a model of FA, we examined the potential of tempol, a nitroxide antioxidant and a superoxide dismutase mimetic, as a tumor-delaying agent for solid tumors. Dietary tempol increased the mean tumor-free survival time of Fancd2(-/-) Trp53(+/-) mice by 27% (P < 0.01), from 308 to 390 days, without changing the overall tumor spectrum. More strikingly, tempol delayed the onset of epithelial tumors and increased the mean epithelial tumor-free survival time by 38% (P < 0.0001), from 312 to 432 days, in Fancd2(-/-) Trp53(+/-) mice. These results show that tempol can significantly delay tumor formation in Fancd2(-/-) Trp53(+/-) mice. Furthermore, tempol treatment did not adversely affect the repopulating ability of FA hematopoietic stem cells. The reduction in oxidative DNA damage in tempol-treated FA fibroblasts and mice suggests that its tumor-delaying function may be attributed to its antioxidant activity.
Sigma-Aldrich
Anti-p53 (Ab-1) (Pantropic) Mouse mAb (PAb421), lyophilized, clone PAb421, Calbiochem®
Pricing and availability is not currently available.