Merck
  • The TMEM240 Protein, Mutated in SCA21, Is Expressed in Purkinje Cells and Synaptic Terminals.
The TMEM240 Protein, Mutated in SCA21, Is Expressed in Purkinje Cells and Synaptic Terminals.
Cerebellum (London, England) (2020-02-01)
Mégane Homa, Anne Loyens, Sabiha Eddarkaoui, Emilie Faivre, Vincent Deramecourt, Claude-Alain Maurage, Luc Buée, Vincent Huin, Bernard Sablonnière
ABSTRACT
A variety of missense mutations and a stop mutation in the gene coding for transmembrane protein 240 (TMEM240) have been reported to be the causative mutations of spinocerebellar ataxia 21 (SCA21). We aimed to investigate the expression of TMEM240 protein in mouse brain at the tissue, cellular, and subcellular levels. Immunofluorescence labeling showed TMEM240 to be expressed in various areas of the brain, with the highest levels in the hippocampus, isocortex, and cerebellum. In the cerebellum, TMEM240 was detected in the deep nuclei and the cerebellar cortex. The protein was expressed in all three layers of the cortex and various cerebellar neurons. TMEM240 was localized to climbing, mossy, and parallel fiber afferents projecting to Purkinje cells, as shown by co-immunostaining with VGLUT1 and VGLUT2. Co-immunostaining with synaptophysin, post-synaptic fractionation, and confirmatory electron microscopy showed TMEM240 to be localized to the post-synaptic side of synapses near the Purkinje-cell soma. Similar results were obtained in human cerebellar sections. These data suggest that TMEM240 may be involved in the organization of the cerebellar network, particularly in synaptic inputs converging on Purkinje cells. This study is the first to describe TMEM240 expression in the normal mouse brain.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-VGluT2 Antibody, serum, from guinea pig
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®