MilliporeSigma
  • Two-stage selection of sequences from a random phage display library delineates both core residues and permitted structural range within an epitope.
Two-stage selection of sequences from a random phage display library delineates both core residues and permitted structural range within an epitope.
Journal of immunological methods (1994-01-03)
R M Miceli, M E DeGraaf, H D Fischer
ABSTRACT
Libraries of random peptides can be screened to identify species which interact with antibodies or receptors. Similarly, maps of native molecular interactions can frequently be deduced by screening a limited set of peptide fragments derived from sequences within a native antigen or ligand. However, the existence of cross-reactive sequences that mimic original epitopes and the limited replaceability of amino acid residues suggest that the sequence space accessible by a receptor can be much broader. Definition of this space is of particular importance where structural information is required for peptidomimetic or drug design. We have used a two-stage selection scheme to expand the sequence space accessible by a phage display library and to define peptide epitopes of the anti-FLAG octapeptide monoclonal M2 antibody. Affinity selection of a primary library of 2 x 10(6) random decapeptides identified a non-contiguous core of three residues in the binding motif Tyr-Lys-Xaa-Xaa-Asp. A second stage library with 2 x 10(7) individual clones bearing the core motif but with the remaining flanking and internal residues re-randomized permitted access to a broader sequence space represented in a library equivalent to several orders of magnitude larger. Data here demonstrate that extended access to binding sequence space permitted by multi-stage screening of phage display libraries can reveal not only essential residues required for ligand binding, but also the ligand structural range permitted within the receptor binding pocket.