Merck
  • Functional domains of Litopenaeus vannamei transglutaminase and their involvement in immunoregulation in shrimp.
Functional domains of Litopenaeus vannamei transglutaminase and their involvement in immunoregulation in shrimp.
Fish & shellfish immunology (2018-07-17)
Zhou Zheng, Wenning Xu, Jude Juventus Aweya, Mingqi Zhong, Shangjie Liu, Jingsheng Lun, Jiehui Chen, Yueling Zhang
ABSTRACT
Shrimps, which mainly rely on their innate immune system to response to infectious pathogens, have clottable proteins as an important component of this system. While transglutaminases (TGase) are found in Litopenaeus vannamei and constitute part of the coagulation system, the specific immune-related roles played by its functional domains in the immunoregulation of shrimp has not been well understood. In the present study, we report that the Ig-like domain of L. vannamei transglutaminase (TGase-C) is the main immune-related domain among the three functional domains, as it had higher bacterial agglutinative activity against Vibrio parahaemolyticus and Streptococcus iniae. Using Co-immunoprecipitation and LC-MS/MS analysis, TGase-C was shown to interact with 474 proteins, of which 52 proteins were annotated to L. vannamei. More than half of the L. vannamei annotated proteins have immune-related functions, including apoptosis. Further analysis using pull-down assay revealed that TGase-C interacted with CAP-3 (a homologue of caspase 3). In addition, siRNA-mediated knockdown of LvTGase significantly (p < 0.01) increased the expression level of LvCAP-3 coupled with a significant (p < 0.01) increase in caspase 3/7 activity, suggesting that probably LvTGase participates in shrimp immune response by modulating the activity of LvCAP-3. These findings thus suggest the Ig-like functional domain of L. vannamei's transglutaminase is the domain that is involved in immunoregulation in shrimp.
Sigma-Aldrich
MISSION® esiRNA, targeting human ALAD