Skip to Content
Merck
Diacylglycerol Kinase Inhibitor I solid

Diacylglycerol Kinase Inhibitor I

Diacylglycerol Kinase Inhibitor I solid
Synonym(s):
6-[2-[4-[(4-Fluorophenyl)phenylmethylene]-1-piperidin­yl]ethyl]-7-methyl-5H-thiazolo-[3,2-a]-pyrimidin-5-one, R59022, Diacylglycerol Kinase Inhibitor I
Empirical Formula (Hill Notation):
C27H26FN3OS
CAS Number:
Molecular Weight:
459.58

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Research Area
Quality Segment