Skip to Content
Merck
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt LiChropur™, ≥98.5% (HPLC)

7-Fluorobenzofurazan-4-sulfonic acid ammonium salt

7-Fluorobenzofurazan-4-sulfonic acid ammonium salt LiChropur™, ≥98.5% (HPLC)
Synonym(s):
4-Fluoro-7-sulfobenzofurazan ammonium salt, 7-Fluorobenz-2,1,3-oxadiazole-4-sulfonic acid ammonium salt, Ammonium 7-fluorobenzofurazan-4-sulfonate, SBD-F
Linear Formula:
C6H3FN2O4S · NH3
CAS Number:
Molecular Weight:
235.19

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing