Skip to Content
Merck
All Photos(1)

Documents

PHR9039

Supelco

Metoprolol EP Impurity G

reference material

Synonym(s):

2-(4-Hydroxyphenyl)ethanol, 4-Hydroxyphenethyl alcohol, Tyrosol, p-HPEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
1859884
MDL number:

grade

reference material

Agency

Not traceable to any primary pharmacopoeia standard

API family

metoprolol

Assay

≥97.0% (GC)

form

powder

shelf life

limited shelf life, expiry date on the label

mp

89-92 °C (lit.)

application(s)

pharmaceutical small molecule

storage temp.

−20°C

SMILES string

OCCc1ccc(O)cc1

InChI

1S/C8H10O2/c9-6-5-7-1-3-8(10)4-2-7/h1-4,9-10H,5-6H2

InChI key

YCCILVSKPBXVIP-UHFFFAOYSA-N

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Antioxidant found in olive oil.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service