Skip to Content
Merck
All Photos(1)

Key Documents

N13586

Toxaphene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-79

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

InChI

1S/C10H8Cl8/c1-4-7(2-11,3-12)9(16)6(14)5(13)8(4,15)10(9,17)18/h5-6H,1-3H2

InChI key

OEJNXTAZZBRGDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjian Lao et al.
Journal of chromatography. A, 1270, 262-268 (2012-11-20)
Determinative methods based on gas chromatography-negative chemical ionization mass spectrometry (GC-NCI/MS) provide improved sensitivity and specificity for toxaphene in environmental samples, but are subject to misidentification due to oxygen reaction in the presence of polychlorinated biphenyls (PCBs). The goal of
Juan José Alava et al.
Environmental toxicology and chemistry, 30(7), 1677-1688 (2011-04-22)
Persistent organic pollutants (POPs) are recognized manmade threats to sea turtle populations, but substantial uncertainty exists surrounding their exposure to contaminants and their sensitivity to toxic effects. This uncertainty creates difficulty for conservation managers to make informed decisions for the
Pim E G Leonards et al.
Integrated environmental assessment and management, 8(3), 523-529 (2011-12-14)
A revised risk assessment for toxaphene was developed, based on the assumption that fish consumers are only exposed to toxaphene residues that differ substantially from technical toxaphene due to environmental degradation and metabolism. In vitro studies confirmed that both technical
M H G Berntssen et al.
Aquatic toxicology (Amsterdam, Netherlands), 112-113, 54-61 (2012-03-01)
The relative feed-to-fish accumulation and possible biotransformation of the nona-chlorinated toxaphene congeners currently included in EU-legislation (CHB-50 and -62) and the octa-chlorinated congeners recommended by the European Food Safety Authority to be included in future surveillance of fish samples (CHB-40
Toxaphene.
Report on carcinogens : carcinogen profiles, 12, 418-420 (2011-08-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service