Skip to Content
Merck
All Photos(1)

Documents

72642

Supelco

2-Methoxyhydroquinone

analytical standard

Synonym(s):

2,5-Dihydroxyanisol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)C6H3(OH)2
CAS Number:
Molecular Weight:
140.14
Beilstein:
2045285
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

88-91 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

COc1cc(O)ccc1O

InChI

1S/C7H8O3/c1-10-7-4-5(8)2-3-6(7)9/h2-4,8-9H,1H3

InChI key

LAQYHRQFABOIFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service