28286-U
Empty Tube with Frit for GERSTEL® TD
glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., pkg of 1 ea
Synonym(s):
Empty Tube for Thermal Desorption
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
material
glass TD tube
feature
fritted
packaging
pkg of 1 ea
technique(s)
active air sampling: suitable
O.D. × I.D. × L
6 mm × 4 mm × 7 in.
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with Gerstel® TDS A & TDS2
General description
Empty Tube with Frit for GERSTEL® TD is used for packing other adsorbents or sample preparation applications.
Legal Information
GERSTEL is a registered trademark of Gerstel GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service