22021
Swagelok® Nut
Swagelok® 102-1, brass, 1/16 in. Swagelok
All Photos(1)
About This Item
Recommended Products
material
brass
packaging
pkg of 10 ea
manufacturer/tradename
Swagelok® 102-1
fitting
1/16 in. Swagelok
compatibility
for use with general purpose Supeltex ferrules
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.
Application
Swagelok® nut, front and back ferrule set maybe used for sampler construction from a single capillary gas chromatographic column, for collection of semi-volatile organic compounds through a diffusion denuder.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
Already Own This Product?
Diffusion denuder for the collection of semivolatile organic compounds.
Environmental Science & Technology, 26 (8), 1551-1555 (1992)
The Manipulation of Air-Sensitive Compounds.
Nature, 217-218 (1986)