Z741961
Cameo™ Syringe Filter
PVDF membrane, pore size 0.22 μm, diam. 30 mm
All Photos(1)
About This Item
Recommended Products
material
PVDF membrane
polypropylene housing
sterility
non-sterile
packaging
pkg of 50 ea
manufacturer/tradename
GVS Life Sciences 3038551
diam.
30 mm
pore size
0.22 μm
General description
Designed to speed up and increase sample volume throughput while reducing back pressure, the hold-up volume is slightly lower due to improved flow channel design and reduced spacing between the supports. The design improvements allow an increase in operating pressure the syringe filters can withstand.
Applications:
Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis
Applications:
Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis
Legal Information
Cameo is a trademark of GVS S.p.A.