Z654388
Consort EV series power supplies
EV243, output0-400 V, 0-300 mA, 0-50 W
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
AC/DC input
230 V AC
description
3 × 4 mm sockets
30 ohms min load resistance
manufacturer/tradename
EV243
availability
available only in EU
parameter
0-40 °C ambient temperature
W × D × H
24 cm × 20 cm × 13 cm
weight
3 kg
output
0-400 V, 0-300 mA, 0-50 W
Looking for similar products? Visit Product Comparison Guide
General description
- Timer 0-99.59 hour
- Volt hours 0-99.99 kVH
- LCD display, 2 × 16 characters
- Ground leakage detection 500 μA
- No load detection, programmable, overload detection
- 3600 values of data logging
- 0-95% non condensing humidity
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service