Skip to Content
Merck
All Photos(1)

Key Documents

V2389

Sigma-Aldrich

Virodhamine hydrochloride

>90% (GC), oil

Synonym(s):

Arachidonic acid-(2-aminoethyl)-ester hydrochloride, O-AEA hydrochloride, O-Arachidonoyl ethanolamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H37NO2 · HCl
CAS Number:
Molecular Weight:
384.00
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>90% (GC)

form

oil

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

protect from light
under inert gas

color

pale yellow

solubility

DMF: soluble ≥20 mg/mL
DMSO: soluble ≥20 mg/mL
ethanol: soluble ≥20 mg/mL

shipped in

dry ice

storage temp.

−70°C

SMILES string

Cl[H].CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OCCN

InChI

1S/C22H37NO2.ClH/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22(24)25-21-20-23;/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-21,23H2,1H3;1H/b7-6-,10-9-,13-12-,16-15-;

InChI key

BNTZVCDZOZGRSZ-XVSDJDOKSA-N

Biochem/physiol Actions

Endogenous cannabinoid; partial CB1 receptor agonist; full CB2 receptor agonist.

Packaging

Air- and light-sensitive

Other Notes

Not stable in solution; use dilutions immediately.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy C Porter et al.
The Journal of pharmacology and experimental therapeutics, 301(3), 1020-1024 (2002-05-23)
The first endocannabinoid, anandamide, was discovered in 1992. Since then, two other endocannabinoid agonists have been identified, 2-arachidonyl glycerol and, more recently, noladin ether. Here, we report the identification and pharmacological characterization of a novel endocannabinoid, virodhamine, with antagonist properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service