Skip to Content
Merck
All Photos(1)

Documents

SRP3019

Sigma-Aldrich

BRAK (CXCL14) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Breast and Kidney-expressed chemokine, CXCL14, NJAC, bolekine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.0-10.0 ng/mL

mol wt

9.4 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CXCL14(9547)

General description

Breast and Kidney-expressed chemokine (BRAK) is a CXC chemokine expressed in normal tissue in the absence of inflammatory stimuli, and infrequently expressed in cancer cell lines. BRAK is known to be a highly selective monocyte chemoattractant. However, main function and receptor selectivity is unknown at this time. BRAK contains the four highly conserved cysteine residues present in CXC chemokines. The sequence of the mature protein consists of 87 amino acid residues, and is approximately 30% homologous to the sequences of MIP-2 alpha and beta. Recombinant human BRAK is a 9.4 kDa protein containing 77 amino acid residues.

Biochem/physiol Actions

Breast and Kidney-expressed chemokine (BRAK) is a CXC chemokine expressed in normal tissue in the absence of inflammatory stimuli, and infrequently expressed in cancer cell lines. Recombinant human BRAK is a 9.4 kDa protein containing 77 amino acid residues.

Sequence

SKCKCSRKGP KIRYSDVKKL EMKPKYPHCE EKMVIITTKS VSRYRGQEHC LHPKLQSTKR FIKWYNAWNE KRRVYEE

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Hromas et al.
Biochemical and biophysical research communications, 255(3), 703-706 (1999-03-02)
Chemokines are a family of related proteins that regulate leukocyte infiltration into inflamed tissue and play important roles in many disease processes. Chemokines are divided into two major groups, CC or CXC, based on their sequence around the amino terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service