Skip to Content
Merck
All Photos(1)

Documents

SML1424

Sigma-Aldrich

Ac-YVAD-pNA

≥98% (HPLC)

Synonym(s):

Caspase-1 substrate (Chromogenic), N-α-acetyl-Tyr-Val-Ala-Asp-p-nitroanilide, N-acetyl-L-tyrosyl-L-valyl-L-alanyl-N-(4-nitrophenyl)-L-α-asparagine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H36N6O10
CAS Number:
Molecular Weight:
628.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

SMILES string

O=C([C@H](CC(O)=O)NC([C@H](C)NC([C@H](C(C)C)NC([C@@H](NC(C)=O)CC1=CC=C(O)C=C1)=O)=O)=O)NC2=CC=C([N+]([O-])=O)C=C2

InChI

1S/C29H36N6O10/c1-15(2)25(34-28(42)22(31-17(4)36)13-18-5-11-21(37)12-6-18)29(43)30-16(3)26(40)33-23(14-24(38)39)27(41)32-19-7-9-20(10-8-19)35(44)45/h5-12,15-16,22-23,25,37H,13-14H2,1-4H3,(H,30,43)(H,31,36)(H,32,41)(H,33,40)(H,34,42)(H,38,39)/t16-,22-,23-,25-/m0/s1

InChI key

YDPNOCSPPGFBPX-XNHCRPTKSA-N

Biochem/physiol Actions

Ac-YVAD-pNA is a very selective chromogenic substrate for caspase-1, a key mediator of inflammatory processes formerly known as interleukin (IL)-1-converting enzyme. Ac-YVAD-pNA is used to determine caspase-1 activity. Cleavage of Ac-YVAD-pNA by caspase-1 releases pNA (p-nitroanilide), which can be quantified spectrophotometrically at 405 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service