Skip to Content
Merck
All Photos(1)

Documents

SML1266

Sigma-Aldrich

Bryostatin 2

≥95% (HPLC)

Synonym(s):

(+)-Bryostatin 2, (1S,3S,5Z,7R,8E,11S,12S,13E,15S,17R,21R,23R,25S)-1,11,21,25-Tetrahydroxy-17-[(1R)-1-hydroxyethyl]-5,13-bis(2-methoxy-2-oxoethylidene)-10,10,26,26-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H66O16
CAS Number:
Molecular Weight:
863.00
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Bugula neritina (sponge)

Quality Level

Assay

≥95% (HPLC)

form

film

color

colorless to light yellow

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C45H66O16/c1-9-10-11-12-13-14-37(49)59-41-29(21-39(51)56-8)20-32-24-35(27(2)46)58-40(52)23-30(47)22-33-25-36(48)43(5,6)44(53,60-33)26-34-18-28(19-38(50)55-7)17-31(57-34)15-16-42(3,4)45(41,54)61-32/h11-16,19,21,27,30-36,41,46-48,53-54H,9-10,17-18,20,22-26H2,1-8H3/b12-11+,14-13+,16-15+,28-19-,29-21+/t27-,30-,31?,32?,33-,34+,35-,36+,41+,44+,45-/m1/s1

InChI key

LIPGUSBNMQRYNL-WZYRAJGLSA-N

Biochem/physiol Actions

Bryostatin 2 is potent protein kinase C activator (Ki = 5.9 nM) and an antagonist to phorbol ester tumor promoter 12-0-tetradecanoylphorbol-13-acetate (TPA). Bryostatin 2 exhibits antineoplastic activity against the murine P388 lymphocytic leukemia and human U937 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service