Skip to Content
Merck

SHC015

MISSION® pLKO.1-puro-UbC-TagFP635 Positive Control Plasmid DNA

Contains a gene encoding TagFP635, under the control of the UbC promoter

Synonym(s):

MISSION® Control Vectors

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

shipped in

dry ice

storage temp.

−20°C

General description

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells respectively. In addition, we recommend producing self-inactivating replication incompetent viral particles in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. The UbC-TagFP635 Control Vector is provided as 10 μg of plasmid DNA in Tris-EDTA (TE) buffer at a concentration of 500 ng/μl.

Application

To see more application data, protocols, vector maps visit sigma.com/shrna.
Silencing of the CMV promoter can be a problem in some cell types. For these cells, the ubiquitin promoter (UbC) can be a viable alternative. The MISSION UbC-TagFP635 Control Vector is a 9226 base pair lentivirus plasmid vector that contains a gene encoding TagFP635, under the control of the UbC promoter. The UbC-TagFP635 Control Vector is useful as a positive control in experiments using the MISSION shRNA library clones for applications in which an alternate promoter is necessary.

Legal Information

Use of this product is subject to one or more license agreements. For details, please see http://sigmaaldrich.com/missionlicense.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
TagFP635 is a trademark of Evrogen Co.


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library