SAB5600226
Anti-Dimethyl-Histone H3 (Lys36) antibody, Rabbit monoclonal
recombinant, expressed in HEK 293 cells, clone RM141, purified immunoglobulin
Synonym(s):
H3K36me2, Histone H3 (di methyl K36)
About This Item
Recommended Products
recombinant
expressed in HEK 293 cells
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
RM141, monoclonal
recombinant monoclonal
form
buffered aqueous glycerol solution
species reactivity
vertebrates
technique(s)
ELISA: 0.2-1 μg/mL
multiplexing: 0.1-0.5 μg/mL
western blot: 0.5— 2 μg/mL
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
dimethylation (Lys36)
Gene Information
human ... H3C1(8350)
Specificity
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service