Skip to Content
Merck
All Photos(3)

Documents

SAB5600215

Sigma-Aldrich

Anti-Human IgE antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM122, purified immunoglobulin

Synonym(s):

IGHE, Immunoglobulin heavy constant epsilon

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM122, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

ELISA: suitable (10ng/well – 100ng/well (for Capture)
0.01ug/mL – 0.1ug/mL (for Detection))

flow cytometry: suitable (assay dependent concentration)
immunocytochemistry: 0.5-2 μg/mL
immunohistochemistry: 0.5—2 μg/mL

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

Specificity

This antibody reacts to human IgE. No cross reactivity with human IgG, IgM, IgD, or IgA.

Immunogen

Human IgE

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service