Skip to Content
Merck
All Photos(1)

Documents

SAB3500466

Sigma-Aldrich

Monoclonal Anti-PUMA antibody produced in mouse

clone 10C5G1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Bbc3, Anti-Bcl-2 binding component 3, Anti-P53 upregulated modulator of apoptosis

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10C5G1, monoclonal

form

buffered aqueous solution

species reactivity

rat, human

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BBC3(27113)

Related Categories

Immunogen

Mouse monoclonal PUMA antibody was raised against a recombinant protein corresponding to amino acids 76 – 170 of human PUMA-alpha.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service